CAS RN 131722-67-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(1R-endo)-8-Methyl-3-oxo-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester (2R,3R)-2,3-Dihydroxybutanedioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1R-endo)-8-Methyl-3-oxo-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester (2R,3R)-2,3-Dihydroxybutanedioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1R-endo)-8-Methyl-3-oxo-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester (2R,3R)-2,3-Dihydroxybutanedioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.