CAS RN 130996-24-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Diethyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-2,6-dimethylpyridine-3,5-dicarboxylate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.