CAS RN 13014-18-1
IUPAC Navn:
2,4-dichloro-1-(trichloromethyl)benzene
Synonymer:
2,4-dichloro-1-(trichloromethyl)benzene
Molekylvægt (g/mol):
264.35
Molekylformel:
C7H3Cl5
InChi Key:
KZSNBJMYJWDVTK-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C(Cl)=C1)C(Cl)(Cl)Cl
1
–
1
af
1
Resultater
1
2,4-Dichlorobenzotrichloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.