CAS RN 1300746-79-5
IUPAC Navn:
λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide
Synonymer:
λ¹-copper(1+) phen trifluoromethanide
Molekylvægt (g/mol):
312.76
Molekylformel:
C13H8CuF3N2
InChi Key:
HFYRAUCEONFIPJ-UHFFFAOYSA-N
SMILES:
[Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
1
–
3
af
3
Resultater
1
(1,10-Phenanthroline)(trifluoromethyl)copper, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1,10-Phenanthroline)(trifluoromethyl)copper, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1,10-Phenanthroline)(trifluoromethyl)copper, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.