CAS RN 128322-44-1
IUPAC Navn:
6-(4-methoxyphenyl)-2-methyl-3H,7H-imidazo[1,2-a]pyrazin-3-one hydrochloride
Synonymer:
6-(4-methoxyphenyl)-2-methyl-7H-imidazo[1,2-a]pyrazin-3-one hydrochloride
Molekylvægt (g/mol):
291.74
Molekylformel:
C14H14ClN3O2
InChi Key:
KGMPQNFFCUDXSO-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=C(C=C1)C1=CN2C(=O)C(C)=NC2=CN1
1
–
1
af
1
Resultater
1
MedChemExpress MCLA hydrochloride
MedChemExpress MCLA hydrochloride is a chemiluminescent reagent which can be used to quantify aqueous concentrations of superoxide.