CAS RN 1268454-23-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Serabelisib, MedChemExpress
MedChemExpress Serabelisib (MLN1117) is a selective p110α inhibitor with an IC50 of 15 nM.