CAS RN 1265908-59-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Pemetrexed L-Glutamic Acid Trisodium Salt (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pemetrexed L-Glutamic Acid Trisodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.