CAS RN 1262769-75-4
IUPAC Navn:
3-bromopyrrolidine hydrobromide
Synonymer:
3-bromopyrrolidine hydrobromide
Molekylvægt (g/mol):
230.93
Molekylformel:
C4H9Br2N
InChi Key:
ZSDJSWROXIUGAE-UHFFFAOYNA-N
SMILES:
Br.BrC1CCNC1
1
–
1
af
1
Resultater
1
3-Bromopyrrolidine hydrobromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.