CAS RN 1261590-48-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Duvelisib (R enantiomer), MedChemExpress
MedChemExpress Duvelisib R enantiomer is a PI3K inhibitor, which is the less active enantiomer of Duvelisib.