accessibility menu, dialog, popup

UserName

NaCH3OHOCH3OO

CAS RN 126-96-5

IUPAC Navn: sodium acetic acid acetate
Synonymer: sodium diacetate
Molekylvægt (g/mol): 142.09
Molekylformel: C4H7NaO4
InChi Key: BHZOKUMUHVTPBX-UHFFFAOYSA-M
SMILES: [Na+].CC(O)=O.CC([O-])=O

 Ingen justeringer