CAS RN 125559-00-4
IUPAC Navn:
2,5-dioxopyrrolidin-1-yl 6-({4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexyl}formamido)hexanoate
Synonymer:
2,5-dioxopyrrolidin-1-yl 6-({4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexyl}formamido)hexanoate
Molekylvægt (g/mol):
447.49
Molekylformel:
C22H29N3O7
InChi Key:
IHVODYOQUSEYJJ-UHFFFAOYSA-N
SMILES:
O=C(CCCCCNC(=O)C1CCC(CN2C(=O)C=CC2=O)CC1)ON1C(=O)CCC1=O
1
–
3
af
3
Resultater
1
N-Succinimidyl 6-[[4-(Maleimidomethyl)cyclohexyl]carboxamido] Caproate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Succinimidyl 6-[[4-(Maleimidomethyl)cyclohexyl]carboxamido] Caproate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Succinimidyl 6-[[4-(Maleimidomethyl)cyclohexyl]carboxamido] Caproate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.