CAS RN 1253202-36-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(2S)-3-(2-Chloro-4-nitro-1H-imidazol-1-yl)-4-methoxybenzoic Acid 2-[[4-(Trifluoromethoxy)phenyl]methoxy]propyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S)-3-(2-Chloro-4-nitro-1H-imidazol-1-yl)-4-methoxybenzoic Acid 2-[[4-(Trifluoromethoxy)phenyl]methoxy]propyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.