CAS RN 125313-60-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
3-[1-(3-Hydroxypropyl)-1H-indol-3-yl]-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-[1-(3-Hydroxypropyl)-1H-indol-3-yl]-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.