CAS RN 1251537-19-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(5S,8E,10E,12R)-12-[[(1,1-Dimethylethyl)diphenylsilyl]oxy]-5-hydroxy-15-(trimethylsilyl)-8,10-Pentadecadiene-6,14-diynoic acid 1-Methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(5S,8E,10E,12R)-12-[[(1,1-Dimethylethyl)diphenylsilyl]oxy]-5-hydroxy-15-(trimethylsilyl)-8,10-Pentadecadiene-6,14-diynoic acid 1-Methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.