CAS RN 125112-77-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(3Alpha,5Beta,6Beta,7Alpha)-3-[(Ethoxycarbonyl)oxy]-6-(formyloxy)-7-hydroxycholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3Alpha,5Beta,6Beta,7Alpha)-3-[(Ethoxycarbonyl)oxy]-6-(formyloxy)-7-hydroxycholan-24-oic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.