CAS RN 1247025-84-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ MitoBoronic acid
Ratiometric mass spectrometry mitochondrial hydrogen peroxide (H2O2) probe