CAS RN 123066-64-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
CRAC intermediate 2, MedChemExpress
MedChemExpress CRAC intermediate 2 is a intermediate compound for CRAC inhibitor synthesis, extracted from patent WO 2013059666A1.