CAS RN 1227939-82-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Ilorasertib, MedChemExpress
MedChemExpress Ilorasertib (ABT-348) is a potent and ATP-competitive multitargeted kinase inhibitor, which inhibits Aurora C, Aurora B, and Aurora A with IC50s of 1 nM, 7 nM, 120 nM, respectively. Ilorasertib also suppresses RET tyrosine kinase, PDGFRβ and Flt1 with IC50s of 7 nM, 3 nM and 32 nM, respectively.