CAS RN 1226851-11-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
LCL521, MedChemExpress
MedChemExpress LCL521 is an acid ceramidase (ACDase) inhibitor. LCL521 also inhibits the lysosomal acid sphingomyelinase (ASMase).