CAS RN 1225580-91-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Acamprosate-d12 Calcium (dipropyl-d12), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Acamprosate-d12 Calcium (di-n-propyl-d12), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
Acamprosate-d12 Calcium, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Acamprosate-d12 Calcium (dipropyl-d12), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.