Få mere at vide
Acamprosate-d12 Calcium (di-n-propyl-d12), CDN
Artikelnummer. 30326457
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326457 0.005 g
30310131 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326457 Leverandør CDN Leverandørnr. CDND73810.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acamprosate-d12 Calcium (dipropyl-d12)
CAS 1225580-91-5
Molekylær formel 2 C5 D6 H4 N O4 S . Ca
Mængde 0.005 g
Synonym 1-Propane-1,1,2,2,3,3-d6-sulfonic acid, 3-(acetylamino)-, calcium salt (2:1), 3-(Acetylamino)-1-propanesulfonic-1,1,2,2,3,3-d6 acid hemicalcium salt, Acamprosate-d6 calcium, Acamprosate-d12 calcium, Acamprosate-D6 calcium (propyl-1,1,2,2,3,3-D6), Bis(N-Acetylhomotaurinate-d6) calcium, N-Acetylhomotaurine-d6 hemicalcium salt
InChI formel InChI=1 S/2C5H11NO4S.Ca/c2*1-5(7)6-3-2-4-11(8,9)10;/h2*2-4H2,1H3,(H,6,7)(H,8,9,10);/q;;+2/p-2/i2*2D2,3D2,4D2;
SMIL [Ca+2 ].[2 H]C([2 H])(NC(=O)C)C([2 H])([2 H])C([2 H])([2 H])S(=O)(=O)[O-].[2 H]C([2 H])(NC(=O)C)C([2 H])([2 H])C([2 H])([2 H])S(=O)(=O)[O-]
IUPAC navn calcium;3-acetamido-1,1,2,2,3,3-hexadeuteriopropane-1-sulfonate
Molekylvægt (g/mol) 412.556
Formel vægt 412.104 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity