CAS RN 1219803-96-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
2-Butoxyethanol-1,1,2,2-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
2-Butoxyethanol-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Butoxyethanol-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-Butoxyethanol-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.