Få mere at vide
2-Butoxyethanol-d4, TRC
Artikelnummer. 30341960
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30341960 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341960 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69289750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Butoxyethanol (1,1,2,2-D4)
CAS 1219803-96-9
Molekylær formel C6 D4 H10 O2
Mængde 50 mg
Synonym 2-Butoxyethanol-d4, 2-Butoxyethanol-1,1,2,2-d4, Ethyleneglycol-d4 Monobutyl Ether, Ethyleneglycol Monobutyl Ether-d4, Ethyleneglycol Monobutyl Ether-D4 (2-hydroxyethyl-D4), 2-Butoxy-1,1,2,2-tetradeuterio-ethanol
InChI formel InChI=1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3/i4D2,6D2
SMIL [2H]C([2H])(O)C([2H])([2H])OCCCC
IUPAC navn 2-butoxy-1,1,2,2-tetradeuterio-ethanol
Molekylvægt (g/mol) 122.2
Formel vægt 122.1245