CAS RN 1217816-92-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
rac-2-Methyl-2,3-epoxy-4-phenyl-4H-pyrano[3,2-c]benzopyran-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
rac-2-Methyl-2,3-epoxy-4-phenyl-4H-pyrano[3,2-c]benzopyran-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.