CAS RN 1217700-81-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
[(1S,2R)-3-[[(4-Nitrophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-phenylmethyl)propyl]carbamic Acid, (3S)-Tetrahydro-3-furan-d4-yl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[(1S,2R)-3-[[(4-Nitrophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-phenylmethyl)propyl]carbamic Acid, (3S)-Tetrahydro-3-furan-d4-yl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.