Få mere at vide
[(1S,2R)-3-[[(4-Nitrophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-phenylmethyl)propyl]carbamic Acid, (3S)-Tetrahydro-3-furan-d4-yl Ester, TRC
Artikelnummer. 30472992
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472992 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472992 Leverandør Toronto Research Chemicals Leverandørnr. TRCN50507710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ((1S,2R)-3-(((4-Nitrophenyl)sulfonyl)(2-methylpropyl)amino-2-hydroxy-1-phenylmethyl)propyl)carbamic Acid','(3S)-Tetrahydro-3-furan-d4-yl Ester
CAS 1217700-81-6
Molekylær formel C25H29D4N3O8S
Mængde 10 mg
InChI formel InChI=1S/C25H33N3O8S/c1-18(2)15-27(37(33,34)22-10-8-20(9-11-22)28(31)32)16-24(29)23(14-19-6-4-3-5-7-19)26-25(30)36-21-12-13-35-17-21/h3-11,18,21,23-24,29H,12-17H2,1-2H3,(H,26,30)/t21-,23-,24+/m0/s1/i13D2,17D2
SMIL [2H]C1([2H])C[C@H](OC(=O)N[C@@H](Cc2ccccc2)[C@H](O)CN(CC(C)C)S(=O)(=O)c3ccc(cc3)[N+](=O)[O-])C([2H])([2H])O1
IUPAC navn [(3S)-2,2,5,5-tetradeuteriooxolan-3-yl] N-[(2S,3R)-3-hydroxy-4-[2-methylpropyl-(4-nitrophenyl)sulfonylamino]-1-phenylbutan-2-yl]carbamate
Molekylvægt (g/mol) 539.63
Formel vægt 539.2239