CAS RN 1217608-72-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(2R)-3-Amino-2-fluoropropanoic Acid-13C3 >90% (up to 10% inorganics), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R)-3-Amino-2-fluoropropanoic Acid-13C3, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.