CAS RN 121062-08-6
IUPAC Navn:
(3S,6S,9R,12S,15S,23S)-9-benzyl-6-{3-[(diaminomethylidene)amino]propyl}-15-[(2S)-2-acetamidohexanamido]-12-[(1H-imidazol-5-yl)methyl]-3-[(1H-indol-3-yl)methyl]-2,5,8,11,14,17-hexaoxo-1,4,7,10,13,18-hexaazacyclotricosane-23-carboxamide
Synonymer:
melanotan II
Molekylvægt (g/mol):
1024.2
Molekylformel:
C50H69N15O9
InChi Key:
JDKLPDJLXHXHNV-MFVUMRCOSA-N
SMILES:
CCCC[C@H](NC(C)=O)C(=O)N[C@H]1CC(=O)NCCCC[C@H](NC(=O)[C@H](CC2=CNC3=CC=CC=C23)NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@@H](CC2=CC=CC=C2)NC(=O)[C@H](CC2=CN=CN2)NC1=O)C(N)=O
1
–
4
af
4
Resultater
1
MedChemExpress Melanotan (MT)-II
MedChemExpress Melanotan (MT)-II, a synthetic melanocortin receptor agonist, is an injectable peptide hormone used to promote tanning.
2
MedChemExpress Melanotan (MT)-II
MedChemExpress Melanotan (MT)-II, a synthetic melanocortin receptor agonist, is an injectable peptide hormone used to promote tanning.
3
MedChemExpress Melanotan (MT)-II
MedChemExpress Melanotan (MT)-II, a synthetic melanocortin receptor agonist, is an injectable peptide hormone used to promote tanning.