CAS RN 1198300-79-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
Cerdulatinib (PRT2070), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cerdulatinib (PRT2070), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Cerdulatinib (PRT2070), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Cerdulatinib, MedChemExpress
MedChemExpress Cerdulatinib (PRT062070) is a selective Tyk2 inhibitor with an IC50 of 0.5 nM. Cerdulatinib (PRT062070) also is a dual JAK and SYK inhibitor with IC50s of 12, 6, 8 and 32 for JAK1, 2, 3 and SYK, respectively.