Få mere at vide
Cerdulatinib, MedChemExpress
Artikelnummer. 16483017
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
Pakningsstørrelse:
100mg
10mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16483017 50 mg -
16477036 100 mg 100mg
16469507 5 mg -
16421216 200 mg -
16416968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16483017 Leverandør Medchem Express Leverandørnr. HY15999
MedChemExpress Cerdulatinib (PRT062070) is a selective Tyk2 inhibitor with an IC50 of 0.5 nM. Cerdulatinib (PRT062070) also is a dual JAK and SYK inhibitor with IC50s of 12, 6, 8 and 32 for JAK1, 2, 3 and SYK, respectively.
Cerdulatinib (PRT062070) is a selective Tyk2 inhibitor with an IC50 of 0.5 nM. Cerdulatinib (PRT062070) also is a dual JAK and SYK inhibitor with IC50s of 12, 6, 8 and 32 for JAK1, 2, 3 and SYK, respectively.
Tekniske data
Kemisk navn eller materiale Cerdulatinib
CAS 1198300-79-6
Farve Earth Yellow
Renhedsgrad noter Research
Molekylær formel C20H27N7O3S
Mængde 50 mg
Synonym PRT062070 PRT2070
Opløselighedsinformation DMSO : ≥ 30 mg/mL (67.33 mM)
SMIL O=S(N(CC1)CCN1C2=CC=C(NC3=NC=C(C(N)=O)C(NC4CC4)=N3)C=C2)(CC)=O
Molekylvægt (g/mol) 445.54
Formel vægt 445.54
Procent renhed 98.25%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease