CAS RN 118525-35-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Sagittatoside A, MedChemExpress
MedChemExpress Sagittatoside A is a natural compound isolated from traditional Chinese herb Yinyanghuo (Herba Epimdii).