CAS RN 1185247-50-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
5-[4-[2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]ethoxy-d4]benzylidene]-2,4-thiazolidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-[4-[2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]ethoxy-d4]benzylidene]-2,4-thiazolidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.