Få mere at vide
5-[4-[2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]ethoxy-d4]benzylidene]-2,4-thiazolidinedione, TRC
Artikelnummer. 30475338
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30475338 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475338 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3036772.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-[4-[2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]ethoxy-d4]benzylidene]-2,4-thiazolidinedione
CAS 1185247-50-0
Molekylær formel C21H16D4N2O5S
Mængde 2.5 mg
Synonym 5-[[4-[2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridinyl]ethoxy-d4]phenyl]methylene]-2,4-thiazolidinedione
InChI formel InChI=1S/C21H20N2O5S/c1-21(27-10-11-28-21)15-4-5-16(22-13-15)8-9-26-17-6-2-14(3-7-17)12-18-19(24)23-20(25)29-18/h2-7,12-13H,8-11H2,1H3,(H,23,24,25)/b18-12-/i8D2,9D2
SMIL [2H]C([2H])(Oc1ccc(\C=C\2/SC(=O)NC2=O)cc1)C([2H])([2H])c3ccc(cn3)C4(C)OCCO4
IUPAC navn (5Z)-5-[[4-[1,1,2,2-tetradeuterio-2-[5-(2-methyl-1,3-dioxolan-2-yl)pyridin-2-yl]ethoxy]phenyl]methylidene]-1,3-thiazolidine-2,4-dione
Molekylvægt (g/mol) 416.48
Formel vægt 416.1344