CAS RN 118399-28-3
IUPAC Navn:
benzyl N-(5-oxooxolan-3-yl)carbamate
Synonymer:
benzyl N-(5-oxooxolan-3-yl)carbamate
Molekylvægt (g/mol):
235.24
Molekylformel:
C12H13NO4
InChi Key:
BNIBNUOPVTZWRT-UHFFFAOYNA-N
SMILES:
O=C(NC1COC(=O)C1)OCC1=CC=CC=C1
1
–
3
af
3
Resultater
1
3(R)-[(Carbobenzyloxy)amino]-gamma-butyrolactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3(R)-[(Carbobenzyloxy)amino]-Gamma-butyrolactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3(R)-[(Carbobenzyloxy)amino]-gamma-butyrolactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.