CAS RN 118134-30-8
IUPAC Navn:
({8-tert-butyl-1,4-dioxaspiro[4.5]decan-2-yl}methyl)(ethyl)propylamine
Synonymer:
prosper spiroxamine impulse
Molekylvægt (g/mol):
297.48
Molekylformel:
C18H35NO2
InChi Key:
PUYXTUJWRLOUCW-UHFFFAOYNA-N
SMILES:
CCCN(CC)CC1COC2(CCC(CC2)C(C)(C)C)O1
1
–
4
af
4
Resultater
1
Spiroxamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Spiroxamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Spiroxamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.