missing translation for 'onlineSavingsMsg'
Få mere at vide

Spiroxamine, TRC

Artikelnummer. 30418237
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418237 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30418237 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6835001G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Spiroxamine
CAS 118134-30-8
Renhedsgrad noter GC
Molekylær formel C18 H35 N O2
Mængde 1 g
Synonym 8-(1,1-Dimethylethyl)-N-ethyl-N-propyl-1,4-dioxaspiro[4.5]decane-2-methanamine, Impulse, Impulse (pesticide), KWG 4168, Prosper, Prosper (fungicide), Prosper Hoggar, Spiroketalamine, Spiroxamine, 1,4-Dioxaspiro[4.5]decane-2-methanamine','8-(1,1-dimethylethyl)-N-ethyl-N-propyl-
InChI formel InChI=1S/C18H35NO2/c1-6-12-19(7-2)13-16-14-20-18(21-16)10-8-15(9-11-18)17(3,4)5/h15-16H,6-14H2,1-5H3
SMIL CCCN(CC)CC1COC2(CCC(CC2)C(C)(C)C)O1
IUPAC navn N-[(8-tert-butyl-1,4-dioxaspiro[4.5]decan-3-yl)methyl]-N-ethylpropan-1-amine
Molekylvægt (g/mol) 297.48
Formel vægt 297.2668
Procent renhed >95
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.