CAS RN 1179507-09-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
1-[4-Bromo-2-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-[4-Bromo-2-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-[4-Bromo-2-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.