CAS RN 1153-07-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Spiro[11H-dibenz[b,e]azepine-11,2'-[1,3]dioxolan]-6(5H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Spiro[11H-dibenz[b,e]azepine-11,2'-[1,3]dioxolan]-6(5H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Spiro[11H-dibenz[b,e]azepine-11,2'-[1,3]dioxolan]-6(5H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.