CAS RN 115287-37-1
IUPAC Navn:
methyl[2-(4-nitrophenoxy)ethyl][2-(4-nitrophenyl)ethyl]amine
Synonymer:
methyl[2-(4-nitrophenoxy)ethyl][2-(4-nitrophenyl)ethyl]amine
Molekylvægt (g/mol):
345.36
Molekylformel:
C17H19N3O5
InChi Key:
BDCVRCLAXLOSLH-UHFFFAOYSA-N
SMILES:
CN(CCOC1=CC=C(C=C1)[N+]([O-])=O)CCC1=CC=C(C=C1)[N+]([O-])=O
1
–
3
af
3
Resultater
1
Methyl-(4-nitrophenylethyl)-(4-nitrophenoxyethyl)amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl-(4-nitrophenylethyl)-(4-nitrophenoxyethyl)amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl-(4-nitrophenylethyl)-(4-nitrophenoxyethyl)amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.