CAS RN 1151863-28-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N-tert-Butoxycarbonyl (BetaR,5S)-Beta-Amino-3-chloro-4,5-dihydro-5-isoxazoleethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-tert-Butoxycarbonyl (BetaR,5S)-Beta-Amino-3-chloro-4,5-dihydro-5-isoxazoleethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.