CAS RN 113276-94-1
IUPAC Navn:
dihydrogen N-[2-(methylamino)ethyl]isoquinoline-5-sulfonamide dichloride
Synonymer:
dihydrogen protein kinase inhibitor H-8 dichloride
Molekylvægt (g/mol):
338.25
Molekylformel:
C12H17Cl2N3O2S
InChi Key:
RJJLZYZEVNCZIW-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CNCCNS(=O)(=O)C1=C2C=CN=CC2=CC=C1
1
–
3
af
3
Resultater
1
N-[2-(Methylamino)ethyl]-5-isoquinolinesulfonamide Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[2-(Methylamino)ethyl]-5-isoquinolinesulfonamide Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-[2-(Methylamino)ethyl]-5-isoquinolinesulfonamide Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.