CAS RN 1127-76-0
IUPAC Navn:
1-ethylnaphthalene
Synonymer:
1-ethylnaphthalene
Molekylvægt (g/mol):
156.23
Molekylformel:
C12H12
InChi Key:
ZMXIYERNXPIYFR-UHFFFAOYSA-N
SMILES:
CCC1=C2C=CC=CC2=CC=C1
1
–
3
af
3
Resultater
1
1-Ethylnaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Ethylnaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Ethylnaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.