CAS RN 1108147-88-1
IUPAC Navn:
1-[1-(1-methylcyclooctyl)piperidin-4-yl]-2-(piperidin-3-yl)-1H-1,3-benzodiazole trihydrochloride
Synonymer:
mcoppb
Molekylvægt (g/mol):
518.01
Molekylformel:
C26H43Cl3N4
InChi Key:
DTIPEVOPCGEULQ-UHFFFAOYNA-N
SMILES:
Cl.Cl.Cl.CC1(CCCCCCC1)N1CCC(CC1)N1C(=NC2=CC=CC=C12)C1CCCNC1
1
–
3
af
3
Resultater
1
MCOPPB Trihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
MCOPPB Trihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
MCOPPB Trihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.