CAS RN 1100617-38-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(3S)-3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-4,5-dihydro-2-benzoxepin-1(3H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.