Learn More
Lenz Laborglasintrumente Gas Washing Bottles Acc. to Drechsel without Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05500514
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10546060
  • 429.00 DKK / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12596
  • Complete with head
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
Specifications
1000 mL
Borosilicate Glass (3.3), Duran Tubing
NS 29/32