Learn More
Lenz Laborglasintrumente Gas Washing Bottles Acc. to Drechsel without Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05500514
Additional Details : Weight : 0.10000kg
Description
- Acc. to DIN 12596
- Complete with head
- Ground joint connection: NS 29/32
- Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
1000 mL | |
Borosilicate Glass (3.3), Duran Tubing | |
NS 29/32 |