Complete Purification
Our revolutionary patented Thermo Scientific™ KingFisher™ magnetic separation technology lets you process virtually any sample from any source as well as providing performance, flexibility and speed.
Thermo Scientific KingFisher Duo
Low- to medium-throughput system consisting of the KingFisher Duo magnetic particle processor instrument, optimized DNA/RNA isolation kits, plastics consumables and BindIt software.
KingFisher Duo Nucleic Acid Purification
View the features of the Thermo Scientific Kingfisher Duo, a low- to medium-throughput system consisting of the KingFisher Duo magnetic particle processor instrument, optimized DNA/RNA isolation kits, plastics consumables and BindIt software.
Thermo Scientific KingFisher Flex
Flexible solution for different types of sample processing needs. With the 24 configuration, customers can raise the processing volume up to 5mL resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with the 96 magnet head.
Thermo Scientific KingFisher Kits
Thermo Scientific™ KingFisher™ Blood DNA Kit
Designed for rapid automated purification of genomic DNA from whole blood samples.
Thermo Scientific™ KingFisher™ Plant DNA Kit
Designed for rapid automated purification of genomic DNA from plant materials.
Thermo Scientific™ KingFisher™ Cell & Tissue DNA Kit
Designed for rapid automated purification of genomic DNA from cell and bacterial cultures and tissues.