Få mere at vide
Zymostenol, TRC
Artikelnummer. 30455673
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455673 100 mg
30453354 10 mg
30453118 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455673 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ701530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 566-97-2
Molekylær formel C27 H46 O
Molekylvægt (g/mol) 386.65
Synonym 5α-Cholest-8-en-3β-ol, 3β-Hydroxy-5α-cholest-8(9)-ene, Dihydrozymosterol, δ8-Cholestenol, (3β,5α)-Cholest-8-en-3-ol
IUPAC navn (3S,5S,10S,13R,14R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](O)C[C@@H]4CC3
Tekniske data
Kemisk navn eller materiale Zymostenol
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-21,23-24,28H,6-17H2,1-5H3/t19-,20+,21+,23-,24+,26+,27-/m1/s1
Formel vægt 386.3549
Procent renhed >95%