Få mere at vide
Zuclopenthixol Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Zuclopenthixol Hydrochloride |
| CAS | 58045-23-1 |
| Analyt- eller komponentnavne | Zuclopenthixol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H25 Cl N2 O S . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-Piperazineethanol, 4-[(3Z)-3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-, hydrochloride (1:2)1-Piperazineethanol, 4-[(3Z)-3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-, dihydrochloride (9CI), 1-Piperazineethanol, 4-[3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-, dihydrochloride, (Z)-, 9H-Thioxanthene, 1-piperazineethanol deriv., cis-Clopenthixol hydrochloride, Zuclopenthixol Hydrochloride, Zuclopenthixol Decanoate Imp. C (EP), Buspirone Imp. G (EP), Buspirone USP Related Compound G, Buspirone USP RC G, Zuclopenthixol Decanoate Impurity C as Dihydrochloride |
| InChI formel | InChI=1S/C22H25ClN2OS.2ClH/c23-17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)27-22)5-3-9-24-10-12-25(13-11-24)14-15-26;;/h1-2,4-8,16,26H,3,9-15H2;2*1H/b18-5-;; |
| Løsningstype | Neat |
| SMIL | Cl.Cl.OCCN1CCN(CC\C=C/2\c3ccccc3Sc4ccc(Cl)cc24)CC1 |
| Vis mere |