Få mere at vide
Zopiclone-d8, TRC
Artikelnummer. 30451684
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451684 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451684 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ7005021MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zopiclone-d8
CAS 1215518-83-4
Renhedsgrad noter HPLC
Molekylær formel C17H9D8ClN6O3
Mængde 1 mg
Synonym 6-(5-Chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methyl-1-piperazine-2,2,3,3,5,5,6,6-d8-carboxylate, 4-Methyl-1-piperazine-2,2,3,3,5,5,6,6-d8-carboxylic acid 6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, Zopiclone-d8, Zopiclone-D8 (piperazine-D8)
InChI formel InChI=1S/C17H17ClN6O3/c1-22-6-8-23(9-7-22)17(26)27-16-14-13(19-4-5-20-14)15(25)24(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3/i6D2,7D2,8D2,9D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4)C1([2H])[2H]
Molekylvægt (g/mol) 396.86
Formel vægt 396.1553
Procent renhed >95