Få mere at vide
Zopiclone-d8 N-Oxide (Mixture of Diastereomers), TRC
Artikelnummer. 30453866
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30453866 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453866 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ70051210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-Zopiclone piperazine-d8 N-Oxide
CAS 1215741-18-6
Renhedsgrad noter HPLC
Molekylær formel C17 D8 H9 Cl N6 O4
Mængde 10 mg
Synonym 1-Piperazine-d8-carboxylic Acid 4-Methyl-4-oxide 6-(5-Chloro-pyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5yl Ester, RP 29753-d8
InChI formel InChI=1S/C17H17ClN6O4/c1-24(27)8-6-22(7-9-24)17(26)28-16-14-13(19-4-5-20-14)15(25)23(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3/i6D2,7D2,8D2,9D2
SMIL [2H]C1([2H])N(C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4)C([2H])([2H])C([2H])([2H])N(=O)(C)C1([2H])[2H]
IUPAC navn [6-(5-chloro-2-pyridyl)-5-oxo-7H-pyrrolo[3,4-b]pyrazin-7-yl] 2,2,3,3,5,5,6,6-octadeuterio-4-methyl-4-oxo-1,4$l^{5}-diazinane-1-carboxylate
Molekylvægt (g/mol) 412.86
Formel vægt 412.1502
Procent renhed >95